ENA238711832 Enamine

2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD00297197
  • Cas Number : 312309-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238...240
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49747
Availability details for
Quantity
  • Availability
  • Add to Cart