ENA238711841 Enamine

2-[(5-bromothiophen-2-yl)formamido]propanoic acid

  • MDL number MFCD09813763
  • Cas Number : 1396964-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49759
Availability details for
Quantity
  • Availability
  • Add to Cart