ENA238711842 Enamine

7-nitro-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

  • MDL number MFCD00448620
  • Cas Number : 55149-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49760
Availability details for
Quantity
  • Availability
  • Add to Cart