ENA238711860 Enamine

methyl 3-amino-4-methylbenzoate hydrochloride

  • MDL number MFCD11226203
  • Cas Number : 383677-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49780
Availability details for
Quantity
  • Availability
  • Add to Cart