ENA238711873 Enamine

2,4-difluoro-N-[(thiophen-2-yl)methyl]aniline

  • MDL number MFCD11145812
  • Cas Number : 1019513-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49796
Availability details for
Quantity
  • Availability
  • Add to Cart