ENA238711875 Enamine

2-(3-bromophenyl)pyrrolidine

  • MDL number MFCD02663522
  • Cas Number : 383127-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49798

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P272,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P333+P313,P337+P313,P362,P363,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-49798
pictograms GHS07
hstatements H302,H315,H317,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart