ENA238711877 Enamine

1-(2,6-dichloro-3-fluorophenyl)ethan-1-amine

  • MDL number MFCD09736706
  • Cas Number : 954257-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49801
Availability details for
Quantity
  • Availability
  • Add to Cart