ENA238711879 Enamine

butyl(methyl)[(piperidin-4-yl)methyl]amine

  • MDL number MFCD09041259
  • Cas Number : 926260-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49803
Availability details for
Quantity
  • Availability
  • Add to Cart