ENA238711880 Enamine

4-(1-phenylethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11201343
  • Cas Number : 642075-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49804
Availability details for
Quantity
  • Availability
  • Add to Cart