ENA238711881 Enamine

2-(butan-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD01896498
  • Cas Number : 13786-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44219
Availability details for
Quantity
  • Availability
  • Add to Cart