ENA238711885 Enamine

5,6-dimethyl-1-[(oxiran-2-yl)methyl]-1H-1,3-benzodiazole

  • MDL number MFCD09802568
  • Cas Number : 1016706-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49809
Availability details for
Quantity
  • Availability
  • Add to Cart