ENA238711888 Enamine

1-butyl-1H-1,3-benzodiazole-2-sulfonic acid

  • MDL number MFCD00964522
  • Cas Number : 300707-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44221

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44221
Availability details for
Quantity
  • Availability
  • Add to Cart