ENA238711893 Enamine

2-[4-methyl-6-oxo-2-(pyridin-2-yl)-1,6-dihydropyrimidin-5-yl]acetic acid

  • MDL number MFCD12197276
  • Cas Number : 1193387-71-1
  • Molecular Weight : 245.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-49824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49824
Availability details for
Quantity
  • Availability
  • Add to Cart