ENA238711898 Enamine

1-(4-chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09741134
  • Cas Number : 113808-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49831
Availability details for
Quantity
  • Availability
  • Add to Cart