ENA238711901 Enamine

2-(furan-2-yl)-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD01238347
  • Cas Number : 37128-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49835

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49835
Availability details for
Quantity
  • Availability
  • Add to Cart