ENA238711902 Enamine

4-methyl-1-phenylpentan-3-one

  • MDL number MFCD00965989
  • Cas Number : 40463-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-49836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49836
Availability details for
Quantity
  • Availability
  • Add to Cart