ENA238711904 Enamine

1-{[3-(trifluoromethyl)phenyl]methyl}piperidin-4-amine

  • MDL number MFCD09944239
  • Cas Number : 66898-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49838
Availability details for
Quantity
  • Availability
  • Add to Cart