ENA238711943 Enamine

4,5,6,7-tetrahydro-1,2-benzoxazole-3-carboxylic acid

  • MDL number MFCD03701438
  • Cas Number : 90005-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49887
Availability details for
Quantity
  • Availability
  • Add to Cart