ENA238711950 Enamine

[2-amino-1-(4-fluorophenyl)ethyl]dimethylamine

  • MDL number MFCD06753109
  • Cas Number : 842954-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49894
Availability details for
Quantity
  • Availability
  • Add to Cart