ENA238711956 Enamine

ethyl 4-[(hydroxyimino)methyl]-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD12197095
  • Cas Number : 1173625-38-1
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-44234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44234
Availability details for
Quantity
  • Availability
  • Add to Cart