ENA238711959 Enamine

(2,2,2-trifluoroethyl)thiourea

  • MDL number MFCD10691447
  • Cas Number : 84545-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49921
Availability details for
Quantity
  • Availability
  • Add to Cart