ENA238711967 Enamine

4-fluoro-N-[(pyridin-2-yl)methyl]aniline

  • MDL number MFCD11058410
  • Cas Number : 940365-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49929
Availability details for
Quantity
  • Availability
  • Add to Cart