ENA238711979 Enamine

1-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}pyrrolidine-2-carboxylic acid

  • MDL number MFCD11100085

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49943
Availability details for
Quantity
  • Availability
  • Add to Cart