ENA238711980 Enamine

1-(piperidin-4-yl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD12912785
  • Cas Number : 1024829-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49944
Availability details for
Quantity
  • Availability
  • Add to Cart