ENA238711984 Enamine

1H-indazol-4-ol

  • MDL number MFCD06858736
  • Cas Number : 81382-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-165
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49951
Availability details for
Quantity
  • Availability
  • Add to Cart