ENA238711985 Enamine

4-fluoro-N-(5-nitro-1,3-thiazol-2-yl)benzamide

  • MDL number MFCD01214158
  • Cas Number : 326-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49952
Availability details for
Quantity
  • Availability
  • Add to Cart