ENA238711989 Enamine

2-(2-bromo-4-fluorophenoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09741115
  • Cas Number : 954563-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44237
Availability details for
Quantity
  • Availability
  • Add to Cart