ENA238711995 Enamine

3-fluoro-4-(4-methylpiperazin-1-yl)benzoic acid hydrochloride

  • MDL number MFCD12912789
  • Cas Number : 948018-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49970
Availability details for
Quantity
  • Availability
  • Add to Cart