ENA238711996 Enamine

1H-indole-3-carboxamide

  • MDL number MFCD00460642
  • Cas Number : 1670-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49971

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49971
Availability details for
Quantity
  • Availability
  • Add to Cart