ENA238712006 Enamine

3-amino-4-(piperidin-1-yl)benzamide

  • MDL number MFCD03274505
  • Cas Number : 31642-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49981
Availability details for
Quantity
  • Availability
  • Add to Cart