ENA238712021 Enamine

2-amino-5-chloro-N-methyl-N-(1-methylpiperidin-4-yl)benzamide hydrochloride

  • MDL number MFCD12197284
  • Cas Number : 1193388-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49998
Availability details for
Quantity
  • Availability
  • Add to Cart