ENA238712035 Enamine

3-methylcyclohexane-1-carbonitrile

  • MDL number MFCD11641473
  • Cas Number : 38857-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-50017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50017
Availability details for
Quantity
  • Availability
  • Add to Cart