ENA238712042 Enamine

4-bromo-1-(4-fluorophenyl)-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD11203683
  • Cas Number : 1152531-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50024
Availability details for
Quantity
  • Availability
  • Add to Cart