ENA238712046 Enamine

1-(4-aminophenyl)-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD09812821
  • Cas Number : 1016728-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50034

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50034
Availability details for
Quantity
  • Availability
  • Add to Cart