ENA238712047 Enamine

4-amino-2-chloro-N-(2-methylpropyl)benzamide

  • MDL number MFCD09044583
  • Cas Number : 926218-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50035
Availability details for
Quantity
  • Availability
  • Add to Cart