ENA238712048 Enamine

3-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)aniline

  • MDL number MFCD11117367
  • Cas Number : 1036512-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50036
Availability details for
Quantity
  • Availability
  • Add to Cart