ENA238712061 Enamine

4-(methoxycarbonyl)-5-methyl-3-propyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD12912795
  • Cas Number : 1193390-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50056
Availability details for
Quantity
  • Availability
  • Add to Cart