ENA238712062 Enamine

1-(2,5-dimethylfuran-3-carbonyl)piperazine hydrochloride

  • MDL number MFCD12912796
  • Cas Number : 1193387-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50059
Availability details for
Quantity
  • Availability
  • Add to Cart