ENA238712066 Enamine

1-(5-methyl-1,2-oxazole-4-carbonyl)piperazine hydrochloride

  • MDL number MFCD12912798
  • Cas Number : 1193387-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50063
Availability details for
Quantity
  • Availability
  • Add to Cart