ENA238712068 Enamine

N-ethyl-3-fluoroaniline hydrochloride

  • MDL number MFCD12912799
  • Cas Number : 1193388-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50065
Availability details for
Quantity
  • Availability
  • Add to Cart