ENA238712072 Enamine

3-(5-methyl-1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD08059833
  • Cas Number : 915924-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50069

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50069
Availability details for
Quantity
  • Availability
  • Add to Cart