ENA238712079 Enamine

methyl[(piperidin-4-yl)methyl](propan-2-yl)amine

  • MDL number MFCD09041262
  • Cas Number : 926192-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50077
Availability details for
Quantity
  • Availability
  • Add to Cart