ENA238712085 Enamine

4-(2,2,2-trifluoroethoxy)butan-1-amine

  • MDL number MFCD09807062
  • Cas Number : 1016719-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50083
Availability details for
Quantity
  • Availability
  • Add to Cart