ENA238712086 Enamine

5-bromo-N-(propan-2-yl)pyridin-2-amine

  • MDL number MFCD11122316
  • Cas Number : 443339-43-3
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50084
Availability details for
Quantity
  • Availability
  • Add to Cart