ENA238712088 Enamine

ethyl (2E)-2-cyano-3-[(3,5-dimethoxyphenyl)amino]prop-2-enoate

  • MDL number MFCD12912800
  • Cas Number : 712307-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50089
Availability details for
Quantity
  • Availability
  • Add to Cart