ENA238712091 Enamine

N-(prop-2-en-1-yl)cyclopentanamine hydrochloride

  • MDL number MFCD12912802
  • Cas Number : 1176911-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50093

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50093
Availability details for
Quantity
  • Availability
  • Add to Cart