ENA238712094 Enamine

9-methyl-1H,2H,3H,3aH,4H,9H,9aH-pyrrolo[3,4-b]quinolin-1-one hydrochloride

  • MDL number MFCD12912803
  • Cas Number : 1212425-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50096
Availability details for
Quantity
  • Availability
  • Add to Cart