ENA238712096 Enamine

4-chloro-5,7-dimethoxyquinoline-3-carbonitrile

  • MDL number MFCD09935343
  • Cas Number : 331662-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50098
Availability details for
Quantity
  • Availability
  • Add to Cart