ENA238712097 Enamine

(2-amino-5-chlorophenyl)(phenyl)methanol

  • MDL number MFCD01004640
  • Cas Number : 7039-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50099
Availability details for
Quantity
  • Availability
  • Add to Cart