ENA238712103 Enamine

4-nitrobenzene-1,3-diol

  • MDL number MFCD04117945
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50105
Availability details for
Quantity
  • Availability
  • Add to Cart